Methyl 4-(bromomethyl)-3-nitrobenzoate 95%
Supplier: Ambeed
|
Danger
|
A158567-1G
A158567-250MG
77011-098EA
146
USD
77011-098
77011-100
Methyl 4-(bromomethyl)-3-nitrobenzoate 95%
Methyl 4-(bromomethyl)-3-nitrobenzoate
Precaución: for Research Use Only
Formula:
C₉H₈BrNO₄ |
MDL Number:
MFCD12031824 CAS Number: 88089-94-5 UN: 3265 ADR: 8,Ⅱ |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...