2,6-Dibromonitrobenzene 97%

Supplier: Ambeed
Warning

A158500-100G A158500-1G A158500-250MG A158500-5G A158500-25G
77028-536EA 542 USD
77028-536 77028-538 77028-540 77028-542 77600-254
2,6-Dibromonitrobenzene 97%
2,6-Dibromonitrobenzene

Precaución: for Research Use Only

Formula: C₆H₃Br₂NO₂
MDL Number: MFCD09998726
CAS Number: 13402-32-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR