5-Bromo-2-nitrobenzonitrile 98%

Supplier: Ambeed
Warning

A158450-10G A158450-1G A158450-250MG A158450-25G A158450-5G
77028-528EA 116.7 USD
77028-528 77028-530 77028-532 77028-534 77671-242
5-Bromo-2-nitrobenzonitrile 98%
5-Bromo-2-nitrobenzonitrile

Precaución: for Research Use Only

Formula: C₇H₃BrN₂O₂
MDL Number: MFCD10699674
CAS Number: 89642-50-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR