2-Ethyl-3-methylbutan-1-ol 95%

Supplier: Ambeed
Warning

A158426-100MG A158426-1G
77028-508EA 33.95 USD
77028-508 77028-510
2-Ethyl-3-methylbutan-1-ol 95%
2-Ethyl-3-methylbutan-1-ol

Precaución: for Research Use Only

Formula: C₇H₁₆O
MW: 116.201 g/mol
MDL Number: MFCD00089635
CAS Number: 32444-34-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR