2,6-Dichloro-3-nitrobenzoic acid 98%

Supplier: Ambeed
Warning

A158356-100G A158356-100MG A158356-10G A158356-1G A158356-250MG A158356-25G A158356-5G
77028-494EA 295 USD
77028-494 77028-496 77028-498 77028-500 77028-502 77028-504 77028-506
2,6-Dichloro-3-nitrobenzoic acid 98%
2,6-Dichloro-3-nitrobenzoic acid

Precaución: for Research Use Only

Formula: C₇H₃Cl₂NO₄
MDL Number: MFCD00075422
CAS Number: 55775-97-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR