3-Phenoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 97%

Supplier: Ambeed
Warning

Synonyms: 5-Phenoxypyridine-3-boronic acid pinacol ester

A158244-1G A158244-250MG
77028-486EA 194.15 USD
77028-486 77028-488
3-Phenoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine 97%
3-Phenoxy-5-(4,4,5,5-tetramethyl-1,3,2-dioxaborolan-2-yl)pyridine

Precaución: for Research Use Only

Formula: C₁₇H₂₀BNO₃
MW: 297.15 g/mol
MDL Number: MFCD11617852
CAS Number: 1309981-45-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR