2,2,2-Trifluoro-1-(pyridin-3-yl)ethane-1,1-diol 95%

Supplier: Ambeed
Warning

A158232-100MG A158232-1G A158232-250MG
77011-078EA 11.67 USD
77011-078 77011-080 77011-082
2,2,2-Trifluoro-1-(pyridin-3-yl)ethane-1,1-diol 95%
2,2,2-Trifluoro-1-(pyridin-3-yl)ethane-1,1-diol

Precaución: for Research Use Only

Formula: C₇H₆F₃NO₂
MW: 193.12 g/mol
MDL Number: MFCD17676630
CAS Number: 33284-19-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR