4-Methyl-2-thiopheneboronic acid 97%

Supplier: Ambeed
Warning

A158170-1G A158170-250MG A158170-25G A158170-5G A158170-100G A158170-10G
77011-074EA 17 USD
77011-074 77011-076 77890-034 77890-035 77890-032 77890-033
4-Methyl-2-thiopheneboronic acid 97%
4-Methyl-2-thiopheneboronic acid

Precaución: for Research Use Only

Formula: C₅H₇BO₂S
MDL Number: MFCD01319040
CAS Number: 162607-15-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR