(S)-3-Amino-2-methylpropanoic acid hydrochloride 95%

Supplier: Ambeed
Warning

A158156-1G A158156-250MG A158156-5G
76984-938EA 81.69 USD
76984-938 76984-940 76984-942
(S)-3-Amino-2-methylpropanoic acid hydrochloride 95%
(S)-3-Amino-2-methylpropanoic acid hydrochloride

Precaución: for Research Use Only

Formula: C₄H₁₀ClNO₂
MW: 139.58 g/mol
MDL Number: MFCD12198195
CAS Number: 925704-45-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR