2-Amino-4-(benzyloxy)-5-methoxybenzoic acid 98%

Supplier: Ambeed
Warning

A157828-1G A157828-250MG A157828-5G
77011-064EA 11 USD
77011-064 77011-066 77890-031
2-Amino-4-(benzyloxy)-5-methoxybenzoic acid 98%
2-Amino-4-(benzyloxy)-5-methoxybenzoic acid

Precaución: for Research Use Only

Formula: C₁₅H₁₅NO₄
MDL Number: MFCD04114355
CAS Number: 155666-33-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR