7-Chloro-1H-indole-2-carboxylic acid 95%

Supplier: Ambeed
Warning

A157825-1G A157825-250MG A157825-5G
77028-468EA 62.6 USD
77028-468 77028-470 77028-472
7-Chloro-1H-indole-2-carboxylic acid 95%
7-Chloro-1H-indole-2-carboxylic acid

Precaución: for Research Use Only

Formula: C₉H₆ClNO₂
MDL Number: MFCD02664428
CAS Number: 28899-75-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR