1-tert-Butyl 4-ethyl azepane-1,4-dicarboxylate 95%

Supplier: Ambeed
Warning

A157798-100MG A157798-250MG
76984-934EA 57.29 USD
76984-934 76984-936
1-tert-Butyl 4-ethyl azepane-1,4-dicarboxylate 95%
1-tert-Butyl 4-ethyl azepane-1,4-dicarboxylate

Precaución: for Research Use Only

Formula: C₁₄H₂₅NO₄
MW: 271.35 g/mol
MDL Number: MFCD18088889
CAS Number: 912444-90-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR