1-(Ethoxycarbonyl)-4-piperidinecarboxylic acid 97%

Supplier: Ambeed
Warning

Synonyms: 1-(Ethoxycarbonyl)-4-piperidinecarboxylic acid, 1-(Ethoxycarbonyl)piperidine-4-carboxylic acid

A157780-100MG A157780-1G A157780-250MG A157780-25G A157780-5G A157780-100G
76997-634EA 5 USD
76997-634 76997-636 76997-638 76997-640 76997-642 77881-235
1-(Ethoxycarbonyl)-4-piperidinecarboxylic acid 97%
1-(Ethoxycarbonyl)-4-piperidinecarboxylic acid

Precaución: for Research Use Only

Formula: C₉H₁₅NO₄
MDL Number: MFCD07339354
CAS Number: 118133-15-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR