Ruxolitinib phosphate 98%

Supplier: Ambeed
Warning

A157761-10MG A157761-25MG A157761-5MG A157761-1MG A157761-100MG
76984-928EA 16.98 USD
76984-928 76984-930 76984-932 77611-510 77683-317
Ruxolitinib phosphate 98%
Ruxolitinib phosphate

Precaución: For research use only.

Formula: C₁₇H₂₁N₆O₄P
MDL Number: MFCD18452860
CAS Number: 1092939-17-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR