trans-4-Methylamino-cyclohexanol 95%
Supplier: Ambeed
|
Warning
|
Synonyms:
trans-4-(Methylamino)cyclohexanol
A157749-1G
A157749-250MG
A157749-5G
77028-462EA
33
USD
77028-462
77028-464
77028-466
trans-4-Methylamino-cyclohexanol 95%
trans-4-Methylamino-cyclohexanol
Precaución: for Research Use Only
Formula:
C₇H₁₅NO MW: 129.2 g/mol |
MDL Number:
MFCD09954372 CAS Number: 22348-44-3 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...