Ethyl 7-chloro-4-hydroxyquinoline-3-carboxylate 95%
Supplier: Ambeed
|
Warning
|
A157673-1G
A157673-5G
76997-630EA
304
USD
76997-630
76997-632
Ethyl 7-chloro-4-hydroxyquinoline-3-carboxylate 95%
Ethyl 7-chloro-4-hydroxyquinoline-3-carboxylate
Precaución: for Research Use Only
Formula:
C₁₂H₁₀ClNO₃ |
MDL Number:
MFCD00438741 CAS Number: 16600-22-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...