2,3-Dichloropyridine 98%

Supplier: Ambeed
Warning

A157654-1000G A157654-100G A157654-10G A157654-500G A157654-5G A157654-25G
77028-452EA 103 USD
77028-452 77028-454 77028-456 77028-458 77028-460 77319-068
2,3-Dichloropyridine 98%
2,3-Dichloropyridine

Precaución: for Research Use Only

Formula: C₅H₃Cl₂N
MDL Number: MFCD00006229
CAS Number: 2402-77-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR