D(-)-2-Phenylglycine ethyl ester hydrochloride 98%

Supplier: Ambeed
Warning

Synonyms: (R)-Ethyl 2-amino-2-phenylacetate hydrochloride, H-D-Phg-OEt.HCl, (R)-(-)-alpha-Aminophenylacetic acid ethyl ester hydrochloride

A157622-100G A157622-10G A157622-25G A157622-500G A157622-5G
76997-620EA 51 USD
76997-620 76997-622 76997-624 76997-626 76997-628
D(-)-2-Phenylglycine ethyl ester hydrochloride 98%
D(-)-2-Phenylglycine ethyl ester hydrochloride

Precaución: for Research Use Only

Formula: C₁₀H₁₃NO₂·HCl
MDL Number: MFCD00052005
CAS Number: 17609-48-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR