4,4'-Dibromo-2,2'-dinitro-1,1'-biphenyl 99%

Supplier: Ambeed
Danger

A157576-1G A157576-5G
76984-922EA 28 USD
76984-922 76984-924
4,4'-Dibromo-2,2'-dinitro-1,1'-biphenyl 99%
4,4'-Dibromo-2,2'-dinitro-1,1'-biphenyl

Precaución: for Research Use Only

Formula: C₁₂H₆Br₂N₂O₄
MDL Number: MFCD14582984
CAS Number: 91371-12-9
UN: 3077
ADR: 9,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR