Ethyl 2,6-difluoro-4-methoxybenzoate 95%

Supplier: Ambeed
Warning

A157575-100MG A157575-1G A157575-250MG
77011-058EA 43.5 USD
77011-058 77011-060 77011-062
Ethyl 2,6-difluoro-4-methoxybenzoate 95%
Ethyl 2,6-difluoro-4-methoxybenzoate

Precaución: for Research Use Only

Formula: C₁₀H₁₀F₂O₃
MW: 216.18 g/mol
MDL Number: MFCD10566176
CAS Number: 1260849-23-7

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR