3-Chloro-5-nitropyridin-4-ol 97%

Supplier: Ambeed
Warning

A157480-1G A157480-5G
76997-616EA 35 USD
76997-616 76997-618
3-Chloro-5-nitropyridin-4-ol 97%
3-Chloro-5-nitropyridin-4-ol

Precaución: for Research Use Only

Formula: C₅H₃ClN₂O₃
MDL Number: MFCD11044290
CAS Number: 31872-64-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR