2-(3-Bromopyridin-2-yl)acetic acid 95%

Supplier: Ambeed
Warning

A157419-1G A157419-250MG A157419-25G A157419-5G
76984-912EA 45.62 USD
76984-912 76984-914 76984-916 76984-918
2-(3-Bromopyridin-2-yl)acetic acid 95%
2-(3-Bromopyridin-2-yl)acetic acid

Precaución: for Research Use Only

Formula: C₇H₆BrNO₂
MW: 216.03 g/mol
MDL Number: MFCD09999978
CAS Number: 698970-84-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR