2-Chloro-4-fluoro-5-nitrobenzaldehyde 96%

Supplier: Ambeed
Warning

A157375-10G A157375-1G A157375-250MG A157375-25G A157375-5G
76984-902EA 49 USD
76984-902 76984-904 76984-906 76984-908 76984-910
2-Chloro-4-fluoro-5-nitrobenzaldehyde 96%
2-Chloro-4-fluoro-5-nitrobenzaldehyde

Precaución: for Research Use Only

Formula: C₇H₃ClFNO₃
MDL Number: MFCD17168722
CAS Number: 99329-85-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR