4-(4,5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxyphenyl)-4,5-dihydro-1H-imidazole-1-carbonyl)piperazin-2-one 99+%

Supplier: Ambeed
Warning

Synonyms: (Rac)-Nutlin-3

A157368-10MG A157368-1MG A157368-25MG A157368-50MG A157368-5MG
77028-444EA 53.05 USD
77028-444 77028-446 77028-448 77028-450 77667-218
4-(4,5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxyphenyl)-4,5-dihydro-1H-imidazole-1-carbonyl)piperazin-2-one 99+%
4-(4,5-Bis(4-chlorophenyl)-2-(2-isopropoxy-4-methoxyphenyl)-4,5-dihydro-1H-imidazole-1-carbonyl)piperazin-2-one

Precaución: for Research Use Only

Formula: C₃₀H₃₀Cl₂N₄O₄
MW: 581.49 g/mol
MDL Number: MFCD07784509
CAS Number: 890090-75-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR