5-(Benzo[d][1,3]dioxol-5-yl)picolinic acid 98%

Supplier: Ambeed
Warning

A157339-100MG A157339-1G A157339-250MG
76984-896EA 152 USD
76984-896 76984-898 76984-900
5-(Benzo[d][1,3]dioxol-5-yl)picolinic acid 98%
5-(Benzo[d][1,3]dioxol-5-yl)picolinic acid

Precaución: for Research Use Only

Formula: C₁₃H₉NO₄
MDL Number: MFCD16817622
CAS Number: 1242339-65-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR