Methyl 4-bromo-2-fluoro-6-methoxybenzoate 95%

Supplier: Ambeed
Warning

A157329-100MG A157329-1G A157329-250MG
77011-046EA 60.48 USD
77011-046 77011-048 77011-050
Methyl 4-bromo-2-fluoro-6-methoxybenzoate 95%
Methyl 4-bromo-2-fluoro-6-methoxybenzoate

Precaución: for Research Use Only

Formula: C₉H₈BrFO₃
MW: 263.06 g/mol
MDL Number: MFCD23707517
CAS Number: 1427415-25-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR