(4-Chloro-3-pyridinyl)methanol 97%

Supplier: Ambeed
Warning

Synonyms: 4-Chloro-3-(hydroxymethyl)pyridine, (4-Chloropyridin-3-yl)methanol

A157294-100MG A157294-1G A157294-1MG A157294-250MG A157294-5G
76997-596EA 18.04 USD
76997-596 76997-598 76997-600 76997-602 76997-604
(4-Chloro-3-pyridinyl)methanol 97%
(4-Chloro-3-pyridinyl)methanol

Precaución: for Research Use Only

Formula: C₆H₆ClNO
MW: 143.57 g/mol
MDL Number: MFCD09743993
CAS Number: 189449-41-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR