5-Bromo-N2-methylpyridine-2,3-diamine 95%

Supplier: Ambeed
Warning

A157204-1G A157204-250MG A157204-100MG
77011-038EA 119 USD
77011-038 77011-040 77890-028
5-Bromo-N2-methylpyridine-2,3-diamine 95%
5-Bromo-N2-methylpyridine-2,3-diamine

Precaución: for Research Use Only

Formula: C₆H₈BrN₃
MDL Number: MFCD07375019
CAS Number: 89415-54-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR