5-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)-4-methylpyrazole-3-carboxylic acid 95%

Supplier: Ambeed
Warning

A157200-100G A157200-10G A157200-1G A157200-250MG A157200-25G A157200-5G
76997-584EA 399.96 USD
76997-584 76997-586 76997-588 76997-590 76997-592 76997-594
5-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)-4-methylpyrazole-3-carboxylic acid 95%
5-(4-Chlorophenyl)-1-(2,4-dichlorophenyl)-4-methylpyrazole-3-carboxylic acid

Precaución: for Research Use Only

Formula: C₁₇H₁₁Cl₃N₂O₂
MW: 381.6400 g/mol
MDL Number: MFCD06411080
CAS Number: 162758-35-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR