6-Fluoro-2,2'-bipyridine 95%

Supplier: Ambeed
Warning

A157157-100MG A157157-1G A157157-250MG
76984-886EA 56 USD
76984-886 76984-888 76984-890
6-Fluoro-2,2'-bipyridine 95%
6-Fluoro-2,2'-bipyridine

Precaución: for Research Use Only

Formula: C₁₀H₇FN₂
MW: 174.174 g/mol
MDL Number: MFCD22576117
CAS Number: 1223063-81-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR