4'-Methyl-3-chloropropiophenone 98%

Supplier: Ambeed
Warning

Synonyms: 3-Chloro-1-(p-tolyl)propan-1-one, 3-Chloro-1-(4-methylphenyl)propan-1-one

A156957-100MG A156957-250MG A156957-1G A156957-5G
76997-568EA 20.16 USD
76997-568 76997-570 77671-216 77671-218
4'-Methyl-3-chloropropiophenone 98%
4'-Methyl-3-chloropropiophenone

Precaución: for Research Use Only

Formula: C₁₀H₁₁ClO
MW: 182.647 g/mol
MDL Number: MFCD00028195
CAS Number: 22422-21-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR