Methylterephthalonitrile 97%

Supplier: Ambeed
Warning

Synonyms: 2-Methylbenzene-1,4-dicarbonitrile, 2-Methylterephthalonitrile

A156945-1G A156945-250MG A156945-25G A156945-5G A156945-10G
77028-430EA 15.92 USD
77028-430 77028-432 77028-434 77028-436 77600-242
Methylterephthalonitrile 97%
Methylterephthalonitrile

Precaución: for Research Use Only

Formula: C₉H₆N₂
MW: 142.157 g/mol
MDL Number: MFCD00119249
CAS Number: 55984-93-5

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR