tert-Butyl-3-iodo-1H-indazole-1-carboxylate 98%

Supplier: Ambeed
Warning

A156940-10G A156940-1G A156940-250MG A156940-5G
77028-422EA 145 USD
77028-422 77028-424 77028-426 77028-428
tert-Butyl-3-iodo-1H-indazole-1-carboxylate 98%
tert-Butyl-3-iodo-1H-indazole-1-carboxylate

Precaución: for Research Use Only

Formula: C₁₂H₁₃IN₂O₂
MDL Number: MFCD09835520
CAS Number: 290368-00-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR