tert-Butyl ((2-(methylsulfonyl)pyrimidin-4-yl)methyl)carbamate 97%

Supplier: Ambeed
Warning

A156820-100MG A156820-1G A156820-250MG
77011-008EA 9.55 USD
77011-008 77011-010 77011-012
tert-Butyl ((2-(methylsulfonyl)pyrimidin-4-yl)methyl)carbamate 97%
tert-Butyl ((2-(methylsulfonyl)pyrimidin-4-yl)methyl)carbamate

Precaución: for Research Use Only

Formula: C₁₁H₁₇N₃O₄S
MW: 287.34 g/mol
MDL Number: MFCD27996634
CAS Number: 1799434-48-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR