6-(Hydroxymethyl)-2,3-dihydro-1H-inden-1-one 98%

Supplier: Ambeed
Warning

A156778-1G A156778-250MG
77011-004EA 315.09 USD
77011-004 77011-006
6-(Hydroxymethyl)-2,3-dihydro-1H-inden-1-one 98%
6-(Hydroxymethyl)-2,3-dihydro-1H-inden-1-one

Precaución: for Research Use Only

Formula: C₁₀H₁₀O₂
MW: 162.18 g/mol
MDL Number: MFCD18207615
CAS Number: 193819-51-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR