2-(7-Methoxynaphthalen-1-yl)ethanamine hydrochloride 97%

Supplier: Ambeed
Warning

A156426-10G A156426-1G A156426-250MG A156426-25G A156426-5G
77028-392EA 132.62 USD
77028-392 77028-394 77028-396 77028-398 77028-400
2-(7-Methoxynaphthalen-1-yl)ethanamine hydrochloride 97%
2-(7-Methoxynaphthalen-1-yl)ethanamine hydrochloride

Precaución: for Research Use Only

Formula: C₁₃H₁₆ClNO
MW: 237.72 g/mol
MDL Number: MFCD13185884
CAS Number: 139525-77-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR