3-Amino-5-bromo-6-methylpyridin-2-ol 95%

Supplier: Ambeed
Warning

A156316-1G A156316-250MG A156316-25G A156316-5G
76984-854EA 21 USD
76984-854 76984-856 76984-858 76984-860
3-Amino-5-bromo-6-methylpyridin-2-ol 95%
3-Amino-5-bromo-6-methylpyridin-2-ol

Precaución: for Research Use Only

Formula: C₆H₇BrN₂O
MDL Number: MFCD08436399
CAS Number: 85216-55-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR