3-Fluoro-4-(trifluoromethoxy)benzoic acid 97%

Supplier: Ambeed
Warning

A156315-1G A156315-5G A156315-25G
77028-388EA 28.65 USD
77028-388 77028-390 77667-206
3-Fluoro-4-(trifluoromethoxy)benzoic acid 97%
3-Fluoro-4-(trifluoromethoxy)benzoic acid

Precaución: for Research Use Only

Formula: C₈H₄F₄O₃
MW: 224.11 g/mol
MDL Number: MFCD06660202
CAS Number: 886498-89-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR