Methyl malonamate 98%

Supplier: Ambeed
Warning

Synonyms: Methyl 3-amino-3-oxopropanoate

A156300-1G A156300-25G A156300-5G
76984-848EA 28.65 USD
76984-848 76984-850 76984-852
Methyl malonamate 98%
Methyl malonamate

Precaución: for Research Use Only

Formula: C₄H₇NO₃
MW: 117.11 g/mol
MDL Number: MFCD00674533
CAS Number: 51513-29-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR