2-Chloro-5-cyanobenzoic acid 95%

Supplier: Ambeed
Warning

A156198-100MG A156198-1G A156198-250MG A156198-50MG
76984-840EA 66 USD
76984-840 76984-842 76984-844 76984-846
2-Chloro-5-cyanobenzoic acid 95%
2-Chloro-5-cyanobenzoic acid

Precaución: for Research Use Only

Formula: C₈H₄ClNO₂
MDL Number: MFCD11036379
CAS Number: 89891-83-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR