Methyl-3-(4-bromophenyl)propanoate 98%

Supplier: Ambeed
Warning

A156137-100G A156137-10G A156137-1G A156137-25G A156137-5G
77028-378EA 162 USD
77028-378 77028-380 77028-382 77028-384 77028-386
Methyl-3-(4-bromophenyl)propanoate 98%
Methyl-3-(4-bromophenyl)propanoate

Precaución: for Research Use Only

Formula: C₁₀H₁₁BrO₂
MDL Number: MFCD09953150
CAS Number: 75567-84-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR