1-(5-Bromo-2-methoxypyridin-3-yl)ethanone 95%

Supplier: Ambeed
Warning

A156071-100MG A156071-250MG A156071-5G
76984-830EA 5 USD
76984-830 76984-832 76984-834
1-(5-Bromo-2-methoxypyridin-3-yl)ethanone 95%
1-(5-Bromo-2-methoxypyridin-3-yl)ethanone

Precaución: for Research Use Only

Formula: C₈H₈BrNO₂
MDL Number: MFCD18256150
CAS Number: 1256811-02-5

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR