4-Amino-2-methylbenzyl Alcohol 97%

Supplier: Ambeed
Warning

Synonyms: (4-Amino-2-methylphenyl)methanol

A156006-1G A156006-250MG A156006-100MG
76984-818EA 133 USD
76984-818 76984-820 77885-269
4-Amino-2-methylbenzyl Alcohol 97%
4-Amino-2-methylbenzyl Alcohol

Precaución: for Research Use Only

Formula: C₈H₁₁NO
MDL Number: MFCD11110504
CAS Number: 63405-88-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR