3-((Dimethylamino)methyl)azetidine dihydrochloride 97%

Supplier: Ambeed
Warning

Synonyms: 1-(Azetidin-3-yl)-N,N-dimethylmethanamine dihydrochloride

A155815-100MG A155815-250MG
76984-814EA 53.05 USD
76984-814 76984-816
3-((Dimethylamino)methyl)azetidine dihydrochloride 97%
3-((Dimethylamino)methyl)azetidine dihydrochloride

Precaución: for Research Use Only

Formula: C₆H₁₄N₂·2ClH
MW: 187.111 g/mol
MDL Number: MFCD09264464
CAS Number: 321890-22-6

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR