3-Methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulfonyl chloride 98%

Supplier: Ambeed
Danger

Synonyms: 3-Methyl-2-oxo-2,3-dihydrobenzo[d]oxazole-6-sulphonyl chloride

A155809-100MG A155809-1G A155809-250MG
76997-530EA 54.11 USD
76997-530 76997-532 76997-534
3-Methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulfonyl chloride 98%
3-Methyl-2-oxo-2,3-dihydro-1,3-benzoxazole-6-sulfonyl chloride

Precaución: for Research Use Only

Formula: C₈H₆ClNO₄S
MW: 247.66 g/mol
MDL Number: MFCD03033702
CAS Number: 62522-63-8
UN: 3261
ADR: 8,Ⅲ

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR