4-Bromo-1-tosyl-1H-pyrrolo[2,3-b]pyridine 96%

Supplier: Ambeed
Warning

Synonyms: 4-Bromo-1(n)-tosyl-7-azaindole

A155769-250MG A155769-5G
77028-366EA 37.14 USD
77028-366 77028-368
4-Bromo-1-tosyl-1H-pyrrolo[2,3-b]pyridine 96%
4-Bromo-1-tosyl-1H-pyrrolo[2,3-b]pyridine

Precaución: for Research Use Only

Formula: C₁₄H₁₁BrN₂O₂S
MW: 351.2200 g/mol
MDL Number: MFCD10575009
CAS Number: 348640-07-3

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR