2-Chloro-5-methylpyridine-3,4-diamine 97%

Supplier: Ambeed
Warning

A155577-100MG A155577-250MG A155577-1G A155577-5G
76984-810EA 40.32 USD
76984-810 76984-812 77592-912 77683-305
2-Chloro-5-methylpyridine-3,4-diamine 97%
2-Chloro-5-methylpyridine-3,4-diamine

Precaución: For research use only.

Formula: C₆H₈ClN₃
MW: 157.60 g/mol
MDL Number: MFCD09953778
CAS Number: 18232-91-2

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR