3-Bromo-5-(bromomethyl)benzonitrile 95%

Supplier: Ambeed
Danger

A155574-1G A155574-250MG
77010-982EA 67 USD
77010-982 77890-017
3-Bromo-5-(bromomethyl)benzonitrile 95%
3-Bromo-5-(bromomethyl)benzonitrile

Precaución: for Research Use Only

Formula: C₈H₅Br₂N
MDL Number: MFCD09261038
CAS Number: 124289-24-3
UN: 3261
ADR: 8,Ⅲ

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR