5-Bromo-6-methyl-1H-indole 97%

Supplier: Ambeed
Warning

A155542-100MG A155542-250MG A155542-5G A155542-1G
77010-976EA 47.75 USD
77010-976 77010-978 77010-980 77549-978
5-Bromo-6-methyl-1H-indole 97%
5-Bromo-6-methyl-1H-indole

Precaución: for Research Use Only

Formula: C₉H₈BrN
MW: 210.07 g/mol
MDL Number: MFCD09026969
CAS Number: 1000343-13-4

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR