Methyl 3-((tert-butoxycarbonyl)amino)propanoate 95%, Ambeed.Inc

Supplier: Ambeed
Warning

A155518-1G A155518-250MG A155518-5G A155518-25G
76984-806EA 43 USD
76984-806 76984-808 77683-303 77885-266
Methyl 3-((tert-butoxycarbonyl)amino)propanoate 95%, Ambeed.Inc
Methyl 3-((tert-butoxycarbonyl)amino)propanoate

Precaución: For research use only.

Formula: C₉H₁₇NO₄
MDL Number: MFCD12198407
CAS Number: 42116-55-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR